Order Entry
France
ContactUsLinkComponent
Pesticide reference standard, Fenitrothion (O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, Fenitrothion (O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA13480000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Fenitrothion (O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA13480000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    122-14-5
  • Description:
    Fenitrothion (O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate) 100 µg/ml in acetonitrile
  • Formula:
    C₉H₁₂NO₅PS
  • Description:
    Fenitrothion (O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate)

Specifications

About this item