Order Entry
France
ContactUsLinkComponent
Potassium standard solution, 10000 mg/L K in 5% hydrochloric acid (from K) standard for AAS and ICP
Potassium standard solution, 10000 mg/L K in 5% hydrochloric acid (from K) standard for AAS and ICP
Catalog # JOBIPLK1-3X
CAS Number:  
Potassium standard solution, 10000 mg/L K in 5% hydrochloric acid (from K) standard for AAS and ICP
Catalog # JOBIPLK1-3X
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Potassium
  • Symbol:
    K

Specifications

About this item

Certifications: Traceable to SRM from NIST