Tin standard solution, 1000 mg/L Sn in nitric acid with hydrofluoric acid (each max. 1%) (from Sn) standard for ICP-MS
Catalog # JOBICLSN2-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1% HNO₃/1% HF
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Tin
- Symbol:Sn
Specifications
About this item
Certifications: Traceable to SRM from NIST