Order Entry
France
ContactUsLinkComponent
Iron standard solution, 10000 mg/L Fe in 5% hydrochloric acid (from Fe) standard for AAS and ICP
Iron standard solution, 10000 mg/L Fe in 5% hydrochloric acid (from Fe) standard for AAS and ICP
Catalog # JOBIPLFE1-3X
CAS Number:  
Iron standard solution, 10000 mg/L Fe in 5% hydrochloric acid (from Fe) standard for AAS and ICP
Catalog # JOBIPLFE1-3X
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

Certifications: Traceable to SRM from NIST