Yttrium standard solution, 10 mg/L Y in dil. nitric acid standard for ICP-MS
Catalog # JOBICLY2-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Yttrium
- Symbol:Y
Specifications
About this item
Certifications: Traceable to SRM from NIST