Terbium standard solution, 10 mg/L Tb in dil. nitric acid (from Tb) standard for ICP-MS
Catalog # JOBICLTB2-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Terbium
- Symbol:Tb
Specifications
About this item
Certifications: Traceable to SRM from NIST