Rhodium standard solution, 10 mg/L Rh in 2% hydrochloric acid (from Rh) standard for ICP-MS
Catalog # JOBICLRH1-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:2% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Rhodium
- Symbol:Rh
Specifications
About this item
Certifications: Traceable to SRM from NIST