Molybdenum standard solution, 10000 mg/L Mo in water (from Mo) standard for AAS and ICP
Catalog # 634-0220
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Certifications: Traceable to SRM from NIST