Germanium standard solution, 10 mg/L Ge in water (de Ge) standard for ICP-MS
Catalog # JOBICLGE9-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Germanium
- Symbol:Ge
Specifications
About this item
Certifications: Traceable to SRM from NIST