Thulium standard solution, 1000 mg/L Tm in dil. nitric acid (from Tm) standard for AAS and ICP
JOBIPLTM2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Thulium
- Symbol:Tm
Traceable to SRM from NIST
: Traceable to SRM from NIST