Zinc standard solution, 1000 mg/L Zn in 2% hydrochloric acid (from Zn) standard for AAS and ICP
Catalog # 634-0309
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HCl
- Pack type:Glass bottle
- Pk:500 mL
- Element:Zinc
- Symbol:Zn
Specifications
About this item
Certifications: Traceable to SRM from NIST