Order Entry
France
ContactUsLinkComponent
Lithium standard solution, 1000 mg/L Li in 2% hydrochloric acid (from Li) standard for AAS and ICP
Lithium standard solution, 1000 mg/L Li in 2% hydrochloric acid (from Li) standard for AAS and ICP
  634-0207
 :  
Lithium standard solution, 1000 mg/L Li in 2% hydrochloric acid (from Li) standard for AAS and ICP
  634-0207
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Lithium
  • Symbol:
    Li

 

 

 : Traceable to SRM from NIST