Order Entry
France
ContactUsLinkComponent
Thallium standard solution, 10000 mg/L Tl in 5% nitric acid (from Tl) standard for AAS and ICP
Thallium standard solution, 10000 mg/L Tl in 5% nitric acid (from Tl) standard for AAS and ICP
  JOBIPLTL2-3Y
 :  
Thallium standard solution, 10000 mg/L Tl in 5% nitric acid (from Tl) standard for AAS and ICP
  JOBIPLTL2-3Y
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Thallium
  • Symbol:
    Tl

 

 

Traceable to SRM from NIST

 : Traceable to SRM from NIST