Order Entry
France
ContactUsLinkComponent
 
Lanthanum standard solution, 10000 mg/L La in 5% nitric acid (from La) standard for AAS and ICP
  634-0206
 :  
Lanthanum standard solution, 10000 mg/L La in 5% nitric acid (from La) standard for AAS and ICP
  634-0206
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lanthanum
  • Symbol:
    La

 

 

 : Traceable to SRM from NIST