Lithium standard solution, 10000 mg/L Li in 5% nitric acid (from Li) standard for AAS and ICP
Catalog # 634-0210
Fournisseur: JOBIN YVON EMISSION
Numéro CAS:
Spécifications
- Concentration:10000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Lithium
- Symbol:Li
Spécifications
À propos de cet article
Certifications: Traceable to SRM from NIST