Indium standard solution, 10 mg/L In in 5% nitric acid (from In) standard for ICP-MS
Catalog # JOBICLIN2-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Indium
- Symbol:In
Specifications
About this item
Certifications: Traceable to SRM from NIST