Order Entry
France
ContactUsLinkComponent
Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce) standard for AAS and ICP
Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce) standard for AAS and ICP
  634-0162
 :  
Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce) standard for AAS and ICP
  634-0162
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Cerium
  • Symbol:
    Ce

 

 

 : Traceable to SRM from NIST