Order Entry
France
ContactUsLinkComponent
 
Sulphuric acid 60%
Catalog # PRLSMC306122.2500
Supplier:  PROLABO CMP
CAS Number:  
Sulphuric acid 60%
Catalog # PRLSMC306122.2500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

About this item

 :
 :98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
EINECS:231-639-5
UN:1830
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.5…1.8 g/cm³ (20 °C)
ADR:8 II