Order Entry
France
ContactUsLinkComponent
 
Titanium standard solution, 1000 mg/L Ti in water (from Ti) standard for ICP-MS
  JOBICLTI9-2Y
 :  
Titanium standard solution, 1000 mg/L Ti in water (from Ti) standard for ICP-MS
  JOBICLTI9-2Y
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Titanium
  • Symbol:
    Ti

 

 

 : Traceable to SRM from NIST