Thallium standard solution, 1000 mg/L Tl in dil. nitric acid (from Tl) standard for AAS and ICP
JOBIPLTL2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Thallium
- Symbol:Tl
: Traceable to SRM from NIST