Order Entry
France
ContactUsLinkComponent
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP
Catalog # JOBIJYICP-SI
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in water with hydrofluoric acid (max. 1%) standard for ICP
Catalog # JOBIJYICP-SI
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item