Phosphorus standard solution, 1,000 mg/L P in water standard for ICP
Catalog # JOBIJYICP-P
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Element:Phosphorus
- Symbol:P