Order Entry
France
ContactUsLinkComponent
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for ICP
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for ICP
  JOBIJYICP-K
 :  
Potassium standard solution, 1,000 mg/L K in dilute nitric acid standard for ICP
  JOBIJYICP-K
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Potassium
  • Symbol:
    K