Iron standard solution, 1000 mg/L Fe in dil. nitric acid (from Fe) standard for ICP-MS
JOBICLFE2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:125 mL
- Element:Iron
- Symbol:Fe
: Traceable to SRM from NIST