Yttrium standard solution, 1000 mg/L Y in 5% nitric acid standard for AAS and ICP
Catalog # 634-0307
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Yttrium
- Symbol:Y
Specifications
About this item
Certifications: Traceable to SRM from NIST