Order Entry
France
ContactUsLinkComponent
Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb) standard for AAS and ICP
Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb) standard for AAS and ICP
Catalog # JOBIPLYB2-2Y
CAS Number:  
Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb) standard for AAS and ICP
Catalog # JOBIPLYB2-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Ytterbium
  • Symbol:
    Yb

Specifications

About this item

Certifications: Traceable to SRM from NIST