Vanadium standard solution, 1000 mg/L V in dil. nitric acid (from V) standard for ICP-MS
JOBICLV2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Vanadium
- Symbol:V
: Traceable to SRM from NIST