Mercury standard solution, 10000 mg/L Hg in 10% nitric acid (from Hg) standard for AAS and ICP
Catalog # 634-0194
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:10% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Mercury
- Symbol:Hg
Specifications
About this item
Certifications: Traceable to SRM from NIST