Order Entry
France
ContactUsLinkComponent
 
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni) standard for ICP-MS
Catalog # JOBICLNI2-2Y
CAS Number:  
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni) standard for ICP-MS
Catalog # JOBICLNI2-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    1
  • Pk Info:
    1× 125 ml
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Certifications: Traceable to SRM from NIST