Order Entry
France
ContactUsLinkComponent
Sodium standard solution, 1000 mg/L Na in dil. nitric acid standard for ICP
Sodium standard solution, 1000 mg/L Na in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-NA
CAS Number:  
Sodium standard solution, 1000 mg/L Na in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-NA
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item