Sodium standard solution, 1000 mg/L Na in dil. nitric acid (from Na) standard for ICP-MS
Catalog # JOBICLNA2-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Sodium
- Symbol:Na
Specifications
About this item
Certifications: Traceable to SRM from NIST