Molybdenum standard solution, 1000 mg/L Mo in water (from Mo) standard for ICP-MS
Catalog # JOBICLMO9-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Certifications: Traceable to SRM from NIST