Order Entry
France
ContactUsLinkComponent
Cobalt standard solution, 1000 mg/L Co in dil. nitric acid (from Co) standard for ICP-MS
Cobalt standard solution, 1000 mg/L Co in dil. nitric acid (from Co) standard for ICP-MS
  JOBICLCO2-2Y
 :  
Cobalt standard solution, 1000 mg/L Co in dil. nitric acid (from Co) standard for ICP-MS
  JOBICLCO2-2Y
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    1
  • Element:
    Cobalt
  • Symbol:
    Co

 

 

 : Traceable to SRM from NIST