Order Entry
France
ContactUsLinkComponent
Sulphur standard solution, 10000 mg/L S in water (from S) standard for AAS and ICP
Sulphur standard solution, 10000 mg/L S in water (from S) standard for AAS and ICP
Catalog # JOBIPLS9-3Y
CAS Number:  
Sulphur standard solution, 10000 mg/L S in water (from S) standard for AAS and ICP
Catalog # JOBIPLS9-3Y
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item

Certifications: Traceable to SRM from NIST