Sulphur standard solution, 1000 mg/L S in water (from S) standard for AAS and ICP
Catalog # 634-0257
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Sulphur
- Symbol:S
Specifications
About this item
Certifications: Traceable to SRM from NIST