Order Entry
France
ContactUsLinkComponent
Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid (from Mg) standard for ICP-MS
Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid (from Mg) standard for ICP-MS
Catalog # JOBICLMG2-2Y
CAS Number:  
Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid (from Mg) standard for ICP-MS
Catalog # JOBICLMG2-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to SRM from NIST