Manganese standard solution, 1000 mg/L Mn in dil. nitric acid (from Mn) standard for ICP-MS
Catalog # JOBICLMN2-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Manganese
- Symbol:Mn
Specifications
About this item
Certifications: Traceable to SRM from NIST