cis-Aconitic acid tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (1Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid
CAS Number:585-84-2
:
Formula:C₆H₆O₆
MDL Number:MFCD00063184
:174.11 g/mol
Melting Pt:115…120 °C
EINECS:209-561-1
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together