Fumaric acid monoethyl ester 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2459-05-4
:
Formula:C₆H₈O₄
MDL Number:MFCD00002699
:144.13 g/mol
Boiling Pt:147 °C (16 mmHg)
Melting Pt:64…69 °C
Storage Temperature:Ambient
EINECS:219-544-7
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:64 to 69 °C
- Titration with NaOH:≥94 %
Specification Test Results
Frequently Bought Together