


Diisopropyl azodicarboxylate 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2446-83-5
:
Formula:C₈H₁₄N₂O₄
MDL Number:MFCD00008875
:202.21 g/mol
Boiling Pt:75 °C (.2 mmHg)
Melting Pt:3…5 °C
Density:1.02 g/cm³
Flash Pt:106 °C
Storage Temperature:Refrigerator
EINECS:219-502-8
UN:3082
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear to hazy yellow to orange to red Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4180 to 1.4220 (20°C, 589 nm)
- GC:≥92 %
Specification Test Results
Frequently Bought Together