Order Entry
France
ContactUsLinkComponent
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15890000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15890000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    298-00-0
  • Description:
    Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane
  • Formula:
    C₈H₁₀NO₅PS
  • Description:
    Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate)

Specifications

About this item