Supelco® PESTANAL® Organic Reference Standard, Parathion solution
Catalog # 45890-2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 μg/ml in cyclohexane
- Pk:2 mL
- Standards:PESTANAL®, analytical standard
- CAS No.:56-38-2
- Description:Parathion solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00036219
- M-Clarity quality level:MQ100
- Synonym(s):Parathion-ethyl solution
- Molecular weight:291,26
- Flash point:–18 °C – Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331