Order Entry
France
Orders LinkContactUsLinkComponent
trans,trans-2,4-Decadienal

trans,trans-2,4-Decadienal

Supplier:   Cayman Chemical
trans,trans-2,4-Decadienal
Supplier:  Cayman Chemical
Certificates
trans,trans-2,4-Decadienal

4 Options Available Below

Product Details & Documents
Synonyms:  Decadienal, (2E,4E)-Deca-2,4-dienal, DDA
  • CAS Number:25152-84-5
  • Formula:C₁₀H₁₆O
  • MDL Number:MFCD00007007
  • Molecular Weight:152.24 g/mol
  • EINECS:246-668-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 ml
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 61,90 €
    61,90 €
    1 * 5 mL
       61,90 €

  • Glass bottle
    10 ml
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 112,00 €
    112,00 €
    1 * 10 mL
       112,00 €

  • Glass bottle
    25 ml
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 261,00 €
    261,00 €
    1 * 25 mL
       261,00 €

  • Glass bottle
    50 ml
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 460,00 €
    460,00 €
    1 * 50 mL
       460,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.