Order Entry
France
ContactUsLinkComponent
7-Methoxy-1-tetralone
7-Methoxy-1-tetralone
Catalog # MOLEM27521042
Supplier:  Molekula
CAS Number:  
7-Methoxy-1-tetralone
Catalog # MOLEM27521042
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :176.22 g/mol
EINECS:229-916-0
CAS Number:6836-19-7
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001696