1,2,3,4-Tetrahydronaphthalen-1-one (1-Tetralone, α-Tetralone)
Référence MOLEM22571232
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:146.19 g/mol
EINECS:208-460-6
CAS Number:529-34-0
Formula:C₁₀H₁₀O
MDL Number:MFCD00001688