
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:529-34-0
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00001688
:146.19 g/mol
Boiling Pt:257 °C
Melting Pt:6…8 °C
Density:1.097
Flash Pt:129 °C (264 °F)
EINECS:208-460-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together