Order Entry
France
ContactUsLinkComponent
Lutetium standard solution, 10,000 mg/l Lu in 5% nitric acid (from Lu2O3), standard for ICP, Specpure®
Lutetium standard solution, 10,000 mg/l Lu in 5% nitric acid (from Lu2O3), standard for ICP, Specpure®
Catalog # 35757.AE
CAS Number:  
Lutetium standard solution, 10,000 mg/l Lu in 5% nitric acid (from Lu2O3), standard for ICP, Specpure®
Catalog # 35757.AE
CAS Number:  

Specifications

  • Pk:
    100 mL

Specifications

About this item

Molecular Formula: Lu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098

Traceable to NIST