Order Entry
France
ContactUsLinkComponent
Quinoline
Quinoline
Catalog # EHERC16709600
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Quinoline
Catalog # EHERC16709600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :129.16 g/mol
Merck Index:13,08160
Boiling Pt:238 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.095 g/cm³ (20 °C)
Flash Pt:107 °C (closed cup)
EINECS:202-051-6
UN:2656
ADR:6.1,III
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:91-22-5
Formula:C₉H₇N
MDL Number:MFCD00006736