Acetonitrile ≥99.5% for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Molecular Weight:41.05 g/mol
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
UN:1648
ADR:3 II
Merck Index:13,00071
EINECS:200-835-2
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Abs.@220nm:<2
- Abs.@254nm:<0.5
- Assay:>99.5 %
- Liquid Chromatography Suitability:Pass
- Residue After Evaporation:<0.005 %
- Titrable Acid:<8 meq/g
- Titrable Base:<0.6 meq/g
- Water (KF):<0.3 %
Specification Test Results
Frequently Bought Together