
Barium chloride 0.05 mol/l (0.1 N) in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:10361-37-2
:
Formula:BaCl₂
MDL Number:MFCD00003445
:208.24 g/mol
Density:1.034 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
:GHS07
Frequently Bought Together